Product Details

SNP ID
rs146183192
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52845540 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTCTTTGCCGCTGAACTTCTCTGC[G/T]TCTCTGCCAGAAATGGTGTTGAAGG
Phenotype
MIM: 603580
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
PCDH8 PubMed Links

Gene Details

Gene
PCDH8
Gene Name
protocadherin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002590.3 2928 Missense Mutation GAA,GAC E908D NP_002581.2
NM_032949.2 2928 Missense Mutation GAA,GAC E811D NP_116567.1

View Full Product Details