Product Details

SNP ID
rs146570814
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:99965387 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATTAATAATAAATTCTAATGTATC[A/G]TCCGCACAACTTCAGGGTTCCCGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927437 PubMed Links

Gene Details

Gene
LOC101927437
Gene Name
uncharacterized LOC101927437
There are no transcripts associated with this gene.

Gene
ZIC5
Gene Name
Zic family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033132.3 2231 Missense Mutation NP_149123.2

View Full Product Details