Product Details

SNP ID
rs149199630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:52377723 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGGGGCTCTGTTCTCTCAATTCC[A/G]GCCACTAATCCTGCCTGGTGATCCC
Phenotype
MIM: 616821
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
THSD1 PubMed Links

Gene Details

Gene
THSD1
Gene Name
thrombospondin type 1 domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018676.3 2792 Silent Mutation NP_061146.1
NM_199263.2 2792 Silent Mutation NP_954872.1

View Full Product Details