Product Details

SNP ID
rs149762079
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:21671742 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GACCTTGAACCTGTGCCAGTGAAAC[A/G]GCAGATTACTTTTATTTATGCATTT
Phenotype
MIM: 600921
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FGF9 PubMed Links

Gene Details

Gene
FGF9
Gene Name
fibroblast growth factor 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002010.2 667 UTR 5 NP_002001.1
XM_011534996.2 667 Intron XP_011533298.1

View Full Product Details