Product Details

SNP ID
rs138925706
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:77407491 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGGTAGTGGGATGGCAGCTACAAA[C/G]CTGTACACAATGCTGGCCTTCTCAA
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
NOXRED1 PubMed Links

Gene Details

Gene
NOXRED1
Gene Name
NADP dependent oxidoreductase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001113475.2 584 Missense Mutation AGC,AGG S168R NP_001106946.1
XM_005267330.3 584 Missense Mutation AGC,AGG S168R XP_005267387.1
XM_011536427.2 584 Missense Mutation AGC,AGG S168R XP_011534729.1
XM_011536428.2 584 Missense Mutation AGC,AGG S168R XP_011534730.1
XM_011536429.2 584 Missense Mutation AGC,AGG S78R XP_011534731.1
XM_017020969.1 584 Missense Mutation AGC,AGG S168R XP_016876458.1
XM_017020970.1 584 Intron XP_016876459.1

View Full Product Details