Product Details

SNP ID
rs140107931
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:19748358 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGGATAAAACCATGTGGCATTCCA[C/T]TGCCTTGTTAAGGAATGAGCAGAAG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
OR4Q3 PubMed Links

Gene Details

Gene
OR4Q3
Gene Name
olfactory receptor family 4 subfamily Q member 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_172194.1 Intron NP_751944.1

View Full Product Details