Product Details

SNP ID
rs140260851
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:58396333 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGCCTCAAAAGGCTGAGGAGCAG[A/G]GCACGCAGGTGCAGTGCTTTCGATC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TOMM20L PubMed Links

Gene Details

Gene
TOMM20L
Gene Name
translocase of outer mitochondrial membrane 20 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_207377.2 226 Missense Mutation AGC,GGC S58G NP_997260.1
XM_011536742.2 226 Missense Mutation AGC,GGC S58G XP_011535044.1
XM_011536743.2 226 Missense Mutation AGC,GGC S58G XP_011535045.1
XM_011536744.2 226 Intron XP_011535046.1

View Full Product Details