Product Details

SNP ID
rs141489673
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:73478769 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGTAGATCTTTGACAAGTGATGAA[C/T]GAACTTTGCTTCGTGTGGTTAGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HEATR4 PubMed Links

Gene Details

Gene
HEATR4
Gene Name
HEAT repeat containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001220484.1 3748 Missense Mutation CAT,CGT H973R NP_001207413.1
NM_203309.2 3748 Missense Mutation CAT,CGT H973R NP_976054.2
XM_006720143.2 3748 Missense Mutation CAT,CGT H973R XP_006720206.1
XM_011536760.2 3748 Missense Mutation CAT,CGT H973R XP_011535062.1
XM_011536761.2 3748 Intron XP_011535063.1
XM_017021289.1 3748 Missense Mutation CAT,CGT H973R XP_016876778.1

View Full Product Details