Product Details

SNP ID
rs143682573
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74494880 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGTTGACTCTGATAGCTTGGCCTT[C/T]GTGAAAGGGGCCCAGGTGGACTTCA
Phenotype
MIM: 615317 MIM: 602091 MIM: 601015
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ISCA2 PubMed Links

Gene Details

Gene
ISCA2
Gene Name
iron-sulfur cluster assembly 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272007.1 400 UTR 3 NP_001258936.1
NM_194279.3 400 Silent Mutation TTC,TTT F115F NP_919255.2
Gene
LTBP2
Gene Name
latent transforming growth factor beta binding protein 2
There are no transcripts associated with this gene.

Gene
NPC2
Gene Name
NPC intracellular cholesterol transporter 2
There are no transcripts associated with this gene.

View Full Product Details