Product Details

SNP ID
rs143978612
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:23469720 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAGGGGTGGGCTTTGCGAAGATGG[C/T]GGCGCTGGTGAGTTTGGTGTGGTTT
Phenotype
MIM: 610777
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NGDN PubMed Links

Gene Details

Gene
NGDN
Gene Name
neuroguidin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042635.1 32 Missense Mutation GCG,GTG A2V NP_001036100.1
NM_015514.1 32 Missense Mutation GCG,GTG A2V NP_056329.1

View Full Product Details