Product Details

SNP ID
rs144352893
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93347282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTGACGCGATATGCCTCTCCTGC[A/G]TGGGCGCTGTCCTGCCCGCCGCCAC
Phenotype
MIM: 616855 MIM: 616884
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
COX8C PubMed Links

Gene Details

Gene
COX8C
Gene Name
cytochrome c oxidase subunit 8C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_182971.2 92 Missense Mutation CAT,CGT H5R NP_892016.1
Gene
UNC79
Gene Name
unc-79 homolog (C. elegans)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020818.3 92 Intron NP_065869.3
XM_011537018.2 92 Intron XP_011535320.1
XM_011537020.2 92 Intron XP_011535322.1
XM_011537021.2 92 Intron XP_011535323.1
XM_011537022.2 92 Intron XP_011535324.1
XM_011537023.2 92 Intron XP_011535325.1
XM_011537024.2 92 Intron XP_011535326.1
XM_011537025.2 92 Intron XP_011535327.1
XM_011537026.2 92 Intron XP_011535328.1
XM_011537027.2 92 Intron XP_011535329.1
XM_011537028.2 92 Intron XP_011535330.1
XM_011537029.2 92 Intron XP_011535331.1
XM_017021506.1 92 Intron XP_016876995.1
XM_017021507.1 92 Intron XP_016876996.1
XM_017021508.1 92 Intron XP_016876997.1
XM_017021509.1 92 Intron XP_016876998.1
XM_017021510.1 92 Intron XP_016876999.1
XM_017021511.1 92 Intron XP_016877000.1
XM_017021512.1 92 Intron XP_016877001.1
XM_017021513.1 92 Intron XP_016877002.1
XM_017021514.1 92 Intron XP_016877003.1
XM_017021515.1 92 Intron XP_016877004.1
XM_017021516.1 92 Intron XP_016877005.1
XM_017021517.1 92 Intron XP_016877006.1
XM_017021518.1 92 Intron XP_016877007.1
XM_017021519.1 92 Intron XP_016877008.1
XM_017021520.1 92 Intron XP_016877009.1

View Full Product Details