Product Details

SNP ID
rs148364724
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:93183781 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTTCAAGCATTGCAGGGCAGG[C/G]TGAAGAGCCTCTAATGCCTGTGCCA
Phenotype
MIM: 609485
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MOAP1 PubMed Links

Gene Details

Gene
MOAP1
Gene Name
modulator of apoptosis 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022151.4 832 Missense Mutation CAC,CAG H154Q NP_071434.2
Gene
TMEM251
Gene Name
transmembrane protein 251
There are no transcripts associated with this gene.

View Full Product Details