Product Details

SNP ID
rs113812701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:43795126 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGTAACAGCCCCAGATAGAGGAG[C/T]ACGCAGGCCCAGCATGAGGCAACCT
Phenotype
MIM: 612784 MIM: 600215 MIM: 605054 MIM: 614550
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HYPK PubMed Links

Gene Details

Gene
HYPK
Gene Name
huntingtin interacting protein K
There are no transcripts associated with this gene.

Gene
MFAP1
Gene Name
microfibrillar associated protein 1
There are no transcripts associated with this gene.

Gene
MIR1282
Gene Name
microRNA 1282
There are no transcripts associated with this gene.

Gene
SERF2
Gene Name
small EDRK-rich factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001018108.3 1371 UTR 3 NP_001018118.1
NM_001199875.1 1371 UTR 3 NP_001186804.1
NM_001199876.1 1371 UTR 3 NP_001186805.1
NM_001199877.1 1371 UTR 3 NP_001186806.1
NM_001199878.1 1371 UTR 3 NP_001186807.1
Gene
SERF2-C15ORF63
Gene Name
SERF2-C15orf63 readthrough
There are no transcripts associated with this gene.

Gene
SERINC4
Gene Name
serine incorporator 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001258031.1 1371 Silent Mutation GTA,GTG V477V NP_001244960.1
NM_001258032.1 1371 Silent Mutation GTA,GTG V233V NP_001244961.1

View Full Product Details