Product Details

SNP ID
rs114644953
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:41183758 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACTTCTTCCATTATTTAACTTA[G/T]TCATTCTCATTCTCCGCATACCAGG
Phenotype
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
EXD1 PubMed Links

Gene Details

Gene
EXD1
Gene Name
exonuclease 3'-5' domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286441.1 1711 UTR 3 NP_001273370.1
NM_152596.3 1711 UTR 3 NP_689809.2
XM_006720415.3 1711 UTR 3 XP_006720478.1
XM_011521298.2 1711 UTR 3 XP_011519600.1
XM_011521299.2 1711 UTR 3 XP_011519601.1
XM_011521300.2 1711 UTR 3 XP_011519602.1
XM_011521301.2 1711 UTR 3 XP_011519603.1
XM_011521302.2 1711 UTR 3 XP_011519604.1
XM_017021970.1 1711 UTR 3 XP_016877459.1
XM_017021971.1 1711 UTR 3 XP_016877460.1

View Full Product Details