Product Details

SNP ID
rs117344392
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:48991265 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGAAGGTGCTCAAGGCTAGGT[C/G]CAAAATAGGTATTTTTTAAAAGGTG
Phenotype
MIM: 615756
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SECISBP2L PubMed Links

Gene Details

Gene
SECISBP2L
Gene Name
SECIS binding protein 2 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193489.1 4549 UTR 3 NP_001180418.1
NM_014701.3 4549 UTR 3 NP_055516.2

View Full Product Details