Product Details

SNP ID
rs140458322
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:90876268 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCTATTGCAGATCTTCGGGGACT[A/G]TTACCACTTCTGGCATCGAGGAGTG
Phenotype
MIM: 190030 MIM: 136950
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FES PubMed Links

Gene Details

Gene
FES
Gene Name
FES proto-oncogene, tyrosine kinase
There are no transcripts associated with this gene.

Gene
FURIN
Gene Name
furin, paired basic amino acid cleaving enzyme
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289823.1 417 Missense Mutation TAT,TGT Y64C NP_001276752.1
NM_001289824.1 417 Missense Mutation TAT,TGT Y64C NP_001276753.1
NM_002569.3 417 Missense Mutation TAT,TGT Y64C NP_002560.1

View Full Product Details