Product Details

SNP ID
rs140817860
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:64156787 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATCTAGCCAGGCTGTCTTGACTG[C/T]CGTGATGAAGAACTGGGAGCCGTTG
Phenotype
MIM: 606274 MIM: 123841
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CSNK1G1 PubMed Links

Gene Details

Gene
CSNK1G1
Gene Name
casein kinase 1 gamma 1
There are no transcripts associated with this gene.

Gene
PPIB
Gene Name
peptidylprolyl isomerase B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000942.4 635 Missense Mutation ACA,GCA T156A NP_000933.1
Gene
SNX22
Gene Name
sorting nexin 22
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024798.2 635 UTR 3 NP_079074.2
XM_005254677.3 635 Intron XP_005254734.1
XM_017022581.1 635 UTR 3 XP_016878070.1

View Full Product Details