Product Details

SNP ID
rs142844685
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76336928 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTATCCTTTTCTGGGTGCTATGGG[G/T]GATCATTCCAAGAGTAAGTATTTCT
Phenotype
MIM: 609481
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ISL2 PubMed Links

Gene Details

Gene
ISL2
Gene Name
ISL LIM homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145805.2 205 Silent Mutation GGG,GGT G15G NP_665804.1
XM_017022505.1 205 Silent Mutation GGG,GGT G15G XP_016877994.1

View Full Product Details