Product Details

SNP ID
rs145221041
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:76338340 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGCGCGGGACAGCGTGTACCACATC[C/G]AGTGCTTCCGCTGCTCCGTGTGCAG
Phenotype
MIM: 609481 MIM: 611611
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ISL2 PubMed Links

Gene Details

Gene
ISL2
Gene Name
ISL LIM homeobox 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145805.2 497 Missense Mutation CAG,GAG Q113E NP_665804.1
XM_017022505.1 497 Missense Mutation CAG,GAG Q113E XP_016877994.1
Gene
SCAPER
Gene Name
S-phase cyclin A associated protein in the ER
There are no transcripts associated with this gene.

View Full Product Details