Product Details

SNP ID
rs145451123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:45402956 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGACGCCCAGTCCCGATCCCGCTG[G/T]GCTGGTCACCCCTCGTACCCGCGTC
Phenotype
MIM: 602360
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GATM PubMed Links

Gene Details

Gene
GATM
Gene Name
glycine amidinotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001321015.1 636 Intron NP_001307944.1
NM_001482.2 636 Intron NP_001473.1
Gene
SPATA5L1
Gene Name
spermatogenesis associated 5 like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001323640.1 636 Missense Mutation GGG,GTG G176V NP_001310569.1
NM_024063.2 636 Missense Mutation GGG,GTG G176V NP_076968.2

View Full Product Details