Product Details

SNP ID
rs145579086
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:56918786 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAGCCGCCTCCCCCTCCCAGACCC[G/T]AGAGCTCGTGCGGGGCAAAGTGAAC
Phenotype
MIM: 600480
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LOC145783 PubMed Links

Gene Details

Gene
LOC145783
Gene Name
uncharacterized LOC145783
There are no transcripts associated with this gene.

Gene
TCF12
Gene Name
transcription factor 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001306219.2 152 Intron NP_001293148.1
NM_001306220.2 152 Intron NP_001293149.1
NM_001322151.1 152 Intron NP_001309080.1
NM_001322152.1 152 UTR 5 NP_001309081.1
NM_001322154.1 152 UTR 5 NP_001309083.1
NM_001322156.1 152 UTR 5 NP_001309085.1
NM_001322157.1 152 UTR 5 NP_001309086.1
NM_001322158.1 152 UTR 5 NP_001309087.1
NM_001322159.1 152 UTR 5 NP_001309088.1
NM_001322161.1 152 Intron NP_001309090.1
NM_001322162.1 152 Intron NP_001309091.1
NM_001322164.1 152 Intron NP_001309093.1
NM_001322165.1 152 Intron NP_001309094.1
NM_003205.3 152 UTR 5 NP_003196.1
NM_207036.1 152 UTR 5 NP_996919.1
NM_207037.1 152 UTR 5 NP_996920.1
NM_207038.1 152 UTR 5 NP_996921.1
NM_207040.1 152 Intron NP_996923.1
XM_011521959.2 152 UTR 5 XP_011520261.1
XM_011521960.2 152 UTR 5 XP_011520262.1
XM_011521961.2 152 UTR 5 XP_011520263.1
XM_011521962.2 152 UTR 5 XP_011520264.1
XM_011521963.2 152 UTR 5 XP_011520265.1
XM_011521965.1 152 Intron XP_011520267.1
XM_011521966.2 152 Intron XP_011520268.1
XM_011521967.1 152 Intron XP_011520269.1
XM_011521969.1 152 Intron XP_011520271.1
XM_017022520.1 152 UTR 5 XP_016878009.1

View Full Product Details