Product Details

SNP ID
rs147018417
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:65124935 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCCAATATATGTCAGCTGGACCCA[C/T]AGCACCCCCTGCAACCATCTTCCAC
Phenotype
MIM: 608138
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDCD7 PubMed Links

Gene Details

Gene
PDCD7
Gene Name
programmed cell death 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005707.1 Intron NP_005698.1

View Full Product Details