Product Details
- SNP ID
-
rs148110342
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.15:90876318 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GACGAAGCGGTCCCTGTCGCCTCAC[C/T]GCCCGCGGCACAGCCGGCTGCAGAG
- Phenotype
-
MIM: 190030
MIM: 136950
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
FES
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs73489557] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- FES
- Gene Name
- FES proto-oncogene, tyrosine kinase
There are no transcripts associated with this gene.
- Gene
- FURIN
- Gene Name
- furin, paired basic amino acid cleaving enzyme
View Full Product Details