Product Details

SNP ID
rs138605816
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:399423 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCGAGCACTACCAGGACCTGCGGAG[A/G]AAGCCCTTCTACCCTGCCCTCATCC
Phenotype
MIM: 615839 MIM: 601818
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DECR2 PubMed Links

Gene Details

Gene
DECR2
Gene Name
2,4-dienoyl-CoA reductase 2, peroxisomal
There are no transcripts associated with this gene.

Gene
LOC100134368
Gene Name
uncharacterized LOC100134368
There are no transcripts associated with this gene.

Gene
NME4
Gene Name
NME/NM23 nucleoside diphosphate kinase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286433.1 301 Intron NP_001273362.1
NM_001286435.1 301 Silent Mutation AGA,AGG R90R NP_001273364.1
NM_001286436.1 301 Silent Mutation AGA,AGG R20R NP_001273365.1
NM_001286438.1 301 Silent Mutation AGA,AGG R20R NP_001273367.1
NM_001286439.1 301 Silent Mutation AGA,AGG R20R NP_001273368.1
NM_001286440.1 301 Silent Mutation AGA,AGG R20R NP_001273369.1
NM_005009.2 301 Silent Mutation AGA,AGG R90R NP_005000.1

View Full Product Details