Product Details

SNP ID
rs139074047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:29859515 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTCATTCCCAGCACACAAGGTGAA[A/C]AGAGCAGGCTGCACACACAGCAAAC
Phenotype
MIM: 605893
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CDIPT PubMed Links

Gene Details

Gene
CDIPT
Gene Name
CDP-diacylglycerol--inositol 3-phosphatidyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286585.1 719 Silent Mutation CTG,CTT L96L NP_001273514.1
NM_001286586.1 719 Silent Mutation CTG,CTT L76L NP_001273515.1
NM_006319.4 719 Silent Mutation CTG,CTT L141L NP_006310.1
XM_005255038.4 719 Silent Mutation CTG,CTT L165L XP_005255095.1
Gene
CDIPT-AS1
Gene Name
CDIPT antisense RNA 1 (head to head)
There are no transcripts associated with this gene.

View Full Product Details