Product Details

SNP ID
rs139092033
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:30444849 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGTTCTGTTGAGGGTAGCCATATT[A/G]AACATGGCTTCCTGATAGGCCAGCT
Phenotype
MIM: 606218
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SEPHS2 PubMed Links

Gene Details

Gene
SEPHS2
Gene Name
selenophosphate synthetase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012248.3 1127 Silent Mutation TTC,TTT F293F NP_036380.2

View Full Product Details