Product Details

SNP ID
rs140337867
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28538819 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCGAGGTAGGAATGGGCCAGGCTA[C/T]AGAGGTCAGATTCATCCAGGCTGGA
Phenotype
MIM: 614812
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
NUPR1 PubMed Links

Gene Details

Gene
NUPR1
Gene Name
nuclear protein 1, transcriptional regulator
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042483.1 356 Missense Mutation TAT,TGT Y30C NP_001035948.1
NM_012385.2 356 Missense Mutation TAT,TGT Y30C NP_036517.1

View Full Product Details