Product Details
- SNP ID
-
rs140470909
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:69109731 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCCTGCGCTCGGCCCAGCGCATCTC[C/T]TTCCCTGACCTCAAGGTGGTCATGG
- Phenotype
-
MIM: 613202
MIM: 602428
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CHTF8
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs2232228] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CHTF8
- Gene Name
- chromosome transmission fidelity factor 8
There are no transcripts associated with this gene.
- Gene
- HAS3
- Gene Name
- hyaluronan synthase 3
View Full Product Details