Product Details

SNP ID
rs140521926
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88699042 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCTCCTGCTGGTCCAGGTTGCGGG[C/T]ACCACAGTACGGGCAGGCGAAGGTG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CTU2 PubMed Links

Gene Details

Gene
CTU2
Gene Name
cytosolic thiouridylase subunit 2
There are no transcripts associated with this gene.

Gene
RNF166
Gene Name
ring finger protein 166
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171815.1 565 Missense Mutation ACC,GCC T76A NP_001165286.1
NM_001171816.1 565 Missense Mutation ACC,GCC T48A NP_001165287.1
NM_178841.3 565 Missense Mutation ACC,GCC T157A NP_849163.1
XM_011522845.2 565 Missense Mutation TAC,TGC Y209C XP_011521147.1
XM_011522846.1 565 Missense Mutation ACC,GCC T157A XP_011521148.1
XM_011522847.1 565 Missense Mutation ACC,GCC T48A XP_011521149.1
XM_017022910.1 565 Missense Mutation TAC,TGC Y235C XP_016878399.1
XM_017022911.1 565 Intron XP_016878400.1
XM_017022912.1 565 Intron XP_016878401.1
XM_017022913.1 565 Missense Mutation ACC,GCC T48A XP_016878402.1

View Full Product Details