Product Details

SNP ID
rs141534499
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:71568789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCACATCGTTCTCAAATTCTGGGAA[A/G]TGTTCCATCTCAATTCCAACCTATA
Phenotype
MIM: 613018
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TAT PubMed Links

Gene Details

Gene
TAT
Gene Name
tyrosine aminotransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000353.2 1245 Silent Mutation CAC,CAT H382H NP_000344.1
Gene
TAT-AS1
Gene Name
TAT antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details