Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_020947.3 | 3440 | UTR 3 | NP_065998.3 | ||
XM_005256074.3 | 3440 | UTR 3 | XP_005256131.1 | ||
XM_005256075.2 | 3440 | UTR 3 | XP_005256132.1 | ||
XM_006721240.2 | 3440 | UTR 3 | XP_006721303.1 | ||
XM_011523248.2 | 3440 | UTR 3 | XP_011521550.1 | ||
XM_011523249.2 | 3440 | Intron | XP_011521551.1 | ||
XM_017023510.1 | 3440 | UTR 3 | XP_016878999.1 | ||
XM_017023511.1 | 3440 | UTR 3 | XP_016879000.1 | ||
XM_017023512.1 | 3440 | UTR 3 | XP_016879001.1 |