Product Details

SNP ID
rs143501486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:2518794 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAGCAGTGGCTTTGCCATCCCCAAG[A/G]CCTTTGGTGGCTTTTCCAGCATGTT
Phenotype
MIM: 108745 MIM: 613577
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AMDHD2 PubMed Links

Gene Details

Gene
AMDHD2
Gene Name
amidohydrolase domain containing 2
There are no transcripts associated with this gene.

Gene
ATP6V0C
Gene Name
ATPase H+ transporting V0 subunit c
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001198569.1 Intron NP_001185498.1
NM_001694.3 Intron NP_001685.1
Gene
TBC1D24
Gene Name
TBC1 domain family member 24
There are no transcripts associated with this gene.

View Full Product Details