Product Details

SNP ID
rs144335469
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25239803 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATGGCTAAGGGGGCATTTAGGAA[A/G]AGAAGATCATCATCAAAAAGTTTTC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ZKSCAN2 PubMed Links

Gene Details

Gene
ZKSCAN2
Gene Name
zinc finger with KRAB and SCAN domains 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012981.4 3324 UTR 3 NP_001012999.3
XM_017023200.1 3324 UTR 3 XP_016878689.1

View Full Product Details