Product Details

SNP ID
rs144797498
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:28502055 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGCATCCTCTCCATGTTGGTCCAG[A/C]GGCCCTGGGTCCCCAGCCCTCCCAG
Phenotype
MIM: 605220 MIM: 607042 MIM: 608273
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
APOBR PubMed Links

Gene Details

Gene
APOBR
Gene Name
apolipoprotein B receptor
There are no transcripts associated with this gene.

Gene
CLN3
Gene Name
CLN3, battenin
There are no transcripts associated with this gene.

Gene
IL27
Gene Name
interleukin 27
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_145659.3 406 Missense Mutation NP_663634.2
XM_011545780.2 406 Missense Mutation XP_011544082.1

View Full Product Details