Product Details

SNP ID
rs144906840
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:11670372 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGCCCGGCAATGCTCATGATGTGCC[C/T]GGTTGAGTTAGGAGGAGGCAGCGTG
Phenotype
MIM: 603032
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SNN PubMed Links

Gene Details

Gene
SNN
Gene Name
stannin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003498.5 Intron NP_003489.1
XM_017023741.1 Intron XP_016879230.1
XM_017023742.1 Intron XP_016879231.1
Gene
TXNDC11
Gene Name
thioredoxin domain containing 11
There are no transcripts associated with this gene.

View Full Product Details