Product Details

SNP ID
rs145367068
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:3088948 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATTACATAGCTGAGGCCAGAAAGC[A/T]ATGCCTCGGCCAGGGAAGGACAGCT
Phenotype
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
ZSCAN10 PubMed Links

Gene Details

Gene
ZSCAN10
Gene Name
zinc finger and SCAN domain containing 10
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001282415.1 2413 UTR 3 NP_001269344.1
NM_001282416.1 2413 UTR 3 NP_001269345.1
NM_032805.2 2413 UTR 3 NP_116194.2
XM_017023791.1 2413 UTR 3 XP_016879280.1
XM_017023792.1 2413 UTR 3 XP_016879281.1

View Full Product Details