Product Details

SNP ID
rs146637306
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:56683184 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTGCAGTTGGCTCCTGTGCCTGTGC[C/T]GGCTCCTGCAAATGCAAAGAGTGCA
Phenotype
MIM: 156355 MIM: 156359
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MT1IP PubMed Links

Gene Details

Gene
MT1IP
Gene Name
metallothionein 1I, pseudogene
There are no transcripts associated with this gene.

Gene
MT1X
Gene Name
metallothionein 1X
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005952.3 Intron NP_005943.1

View Full Product Details