Product Details

SNP ID
rs146742423
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1945891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAGCAATACAACCCTTCAGCATGA[C/T]GAAGTCGTTGTTCACTTCCCCGTAG
Phenotype
MIM: 606216
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MSRB1 PubMed Links

Gene Details

Gene
MSRB1
Gene Name
methionine sulfoxide reductase B1
There are no transcripts associated with this gene.

Gene
RPL3L
Gene Name
ribosomal protein L3 like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005061.2 1498 Missense Mutation NP_005052.1
XM_011522571.2 1498 Missense Mutation XP_011520873.1

View Full Product Details