Product Details
- SNP ID
-
rs147112593
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:89561660 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AGGCGGAACAAGTCCACGGAGTCCC[G/T]GCAGGCCAACGTGCAGCGGCTGAAG
- Phenotype
-
MIM: 113703
MIM: 602783
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RPL13
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs9930567] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RPL13
- Gene Name
- ribosomal protein L13
- Gene
- SNORD68
- Gene Name
- small nucleolar RNA, C/D box 68
There are no transcripts associated with this gene.
- Gene
- SPG7
- Gene Name
- SPG7, paraplegin matrix AAA peptidase subunit
There are no transcripts associated with this gene.
View Full Product Details