Product Details

SNP ID
rs147327212
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:88697610 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACAGGGCCAGAGCAGCCTGGAAGGC[A/G]GCCTCCTCGTCAATACTGTAGTCCT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CTU2 PubMed Links

Gene Details

Gene
CTU2
Gene Name
cytosolic thiouridylase subunit 2
There are no transcripts associated with this gene.

Gene
RNF166
Gene Name
ring finger protein 166
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171815.1 768 Silent Mutation GCC,GCT A143A NP_001165286.1
NM_001171816.1 768 Silent Mutation GCC,GCT A115A NP_001165287.1
NM_178841.3 768 Silent Mutation GCC,GCT A224A NP_849163.1
XM_011522845.2 768 Silent Mutation GCC,GCT A337A XP_011521147.1
XM_011522846.1 768 UTR 3 XP_011521148.1
XM_011522847.1 768 UTR 3 XP_011521149.1
XM_017022910.1 768 Silent Mutation GCC,GCT A363A XP_016878399.1
XM_017022911.1 768 Intron XP_016878400.1
XM_017022912.1 768 Intron XP_016878401.1
XM_017022913.1 768 Silent Mutation GCC,GCT A115A XP_016878402.1

View Full Product Details