Product Details

SNP ID
rs148778202
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:69109671 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGGCTCGGTGGCACTGTGCATTGC[C/T]GCATACCAGGAGGACCCTGACTACT
Phenotype
MIM: 613202 MIM: 602428
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHTF8 PubMed Links

Gene Details

Gene
CHTF8
Gene Name
chromosome transmission fidelity factor 8
There are no transcripts associated with this gene.

Gene
HAS3
Gene Name
hyaluronan synthase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199280.1 367 Silent Mutation GCC,GCT A92A NP_001186209.1
NM_005329.2 367 Silent Mutation GCC,GCT A92A NP_005320.2
NM_138612.2 367 Silent Mutation GCC,GCT A92A NP_619515.1
XM_005255921.2 367 Silent Mutation GCC,GCT A92A XP_005255978.1
XM_011523061.2 367 Silent Mutation GCC,GCT A92A XP_011521363.1

View Full Product Details