Product Details

SNP ID
rs149136295
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:19718319 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAACCGCGGCCATGGCGGCACCG[C/T]GGCAAATCCCCAGCCACATAGTGCG
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IQCK PubMed Links

Gene Details

Gene
IQCK
Gene Name
IQ motif containing K
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001305121.1 712 UTR 5 NP_001292050.1
NM_001305122.1 712 Missense Mutation CGG,TGG R5W NP_001292051.1
NM_153208.2 712 Missense Mutation CGG,TGG R5W NP_694940.1
XM_011545736.2 712 Intron XP_011544038.1
XM_017022933.1 712 Missense Mutation CGG,TGG R5W XP_016878422.1
Gene
KNOP1
Gene Name
lysine rich nucleolar protein 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001012991.2 712 Intron NP_001013009.2
XM_005255317.4 712 Intron XP_005255374.1
XM_006721046.3 712 Intron XP_006721109.1
XM_011545855.1 712 Intron XP_011544157.1
XM_017023231.1 712 Intron XP_016878720.1
XM_017023232.1 712 Intron XP_016878721.1

View Full Product Details