Product Details

SNP ID
rs149183320
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:25217230 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGTGTGAGCCTGAATTTGGCAATGA[C/T]AAGGCCAGGGAGCCGAGCGTGGGTG
Phenotype
MIM: 603750
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AQP8 PubMed Links

Gene Details

Gene
AQP8
Gene Name
aquaporin 8
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001169.2 127 Silent Mutation GAC,GAT D15D NP_001160.2
XM_011545822.2 127 Silent Mutation GAC,GAT D16D XP_011544124.1
XM_011545823.2 127 Silent Mutation GAC,GAT D16D XP_011544125.1

View Full Product Details