Product Details

SNP ID
rs149525759
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:1420328 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTGGCACCTCTCCCCACACCTGT[C/T]TCAACCACTGGTCCGCCTCGCTCTC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C16orf91 PubMed Links

Gene Details

Gene
C16orf91
Gene Name
chromosome 16 open reading frame 91
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001272051.1 394 Missense Mutation AAA,AGA K106R NP_001258980.1
Gene
LOC105371045
Gene Name
uncharacterized LOC105371045
There are no transcripts associated with this gene.

Gene
UNKL
Gene Name
unkempt family like zinc finger
There are no transcripts associated with this gene.

View Full Product Details