Product Details
- SNP ID
-
rs137881249
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:41487001 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCGGTAGGTAGCGATCTCGCCCTCC[A/T]GCCGGGCCTTGACGTCCAGTAACAC
- Phenotype
-
MIM: 602764
MIM: 604540
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT35
PubMed Links
Gene Details
- Gene
- KRT35
- Gene Name
- keratin 35
There are no transcripts associated with this gene.
- Gene
- KRT36
- Gene Name
- keratin 36
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_003771.4 |
1157 |
Missense Mutation |
CAG,CTG |
Q386L |
NP_003762.1 |
View Full Product Details