Product Details
- SNP ID
-
rs138132734
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:81904917 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAAGGCCAGCTCCTTGGCTTCCTT[C/T]TTCTGGTTTCGCGCCTCATACTCCA
- Phenotype
-
MIM: 614534
MIM: 607996
MIM: 602679
MIM: 606212
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ANAPC11
PubMed Links
Gene Details
- Gene
- ANAPC11
- Gene Name
- anaphase promoting complex subunit 11
There are no transcripts associated with this gene.
- Gene
- NPB
- Gene Name
- neuropeptide B
There are no transcripts associated with this gene.
- Gene
- PCYT2
- Gene Name
- phosphate cytidylyltransferase 2, ethanolamine
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184917.2 |
1492 |
Silent Mutation |
AAA,AAG |
K380K |
NP_001171846.1 |
NM_001256433.2 |
1492 |
Silent Mutation |
AAA,AAG |
K308K |
NP_001243362.1 |
NM_001256434.2 |
1492 |
Silent Mutation |
AAA,AAG |
K321K |
NP_001243363.1 |
NM_001256435.2 |
1492 |
Silent Mutation |
AAA,AAG |
K284K |
NP_001243364.1 |
NM_001282203.1 |
1492 |
Silent Mutation |
AAA,AAG |
K284K |
NP_001269132.1 |
NM_001282204.1 |
1492 |
Silent Mutation |
AAA,AAG |
K330K |
NP_001269133.1 |
NM_002861.4 |
1492 |
Silent Mutation |
AAA,AAG |
K362K |
NP_002852.1 |
XM_005256386.3 |
1492 |
Silent Mutation |
AAA,AAG |
K348K |
XP_005256443.1 |
XM_005256387.3 |
1492 |
Silent Mutation |
AAA,AAG |
K348K |
XP_005256444.1 |
XM_006722287.3 |
1492 |
Silent Mutation |
AAA,AAG |
K358K |
XP_006722350.1 |
XM_017024910.1 |
1492 |
Silent Mutation |
AAA,AAG |
K340K |
XP_016880399.1 |
XM_017024911.1 |
1492 |
Silent Mutation |
AAA,AAG |
K330K |
XP_016880400.1 |
XM_017024912.1 |
1492 |
Silent Mutation |
AAA,AAG |
K193K |
XP_016880401.1 |
- Gene
- SIRT7
- Gene Name
- sirtuin 7
There are no transcripts associated with this gene.
View Full Product Details