Product Details

SNP ID
rs138198807
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:48577011 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGCTTGGGCTCCCCGCCGGCGTAA[C/T]TGGGGTTTACTGGACACACACGGAG
Phenotype
MIM: 142966 MIM: 142965 MIM: 610173
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
HOXB3 PubMed Links

Gene Details

Gene
HOXB3
Gene Name
homeobox B3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002146.4 529 Intron NP_002137.4
XM_005257277.3 529 Intron XP_005257334.1
XM_005257280.3 529 Intron XP_005257337.1
XM_005257282.3 529 Intron XP_005257339.1
XM_006721854.2 529 Intron XP_006721917.1
XM_011524707.2 529 Intron XP_011523009.1
XM_011524708.2 529 Intron XP_011523010.1
XM_011524710.1 529 Intron XP_011523012.1
XM_011524719.1 529 Intron XP_011523021.1
XM_011524720.2 529 Intron XP_011523022.1
XM_011524721.2 529 Intron XP_011523023.1
XM_011524726.2 529 Intron XP_011523028.1
XM_017024557.1 529 Intron XP_016880046.1
XM_017024558.1 529 Intron XP_016880047.1
XM_017024559.1 529 Intron XP_016880048.1
XM_017024560.1 529 Intron XP_016880049.1
XM_017024561.1 529 Intron XP_016880050.1
XM_017024562.1 529 Intron XP_016880051.1
XM_017024563.1 529 Intron XP_016880052.1
Gene
HOXB4
Gene Name
homeobox B4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024015.4 529 Missense Mutation AAT,AGT N156S NP_076920.1
Gene
MIR10A
Gene Name
microRNA 10a
There are no transcripts associated with this gene.

View Full Product Details