Product Details

SNP ID
rs138373624
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:4740764 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCATAGTGTGGGCATCAGCCTGGG[A/G]CAAGGGGCAGAACCAGGTCCTGGAC
Phenotype
MIM: 605398 MIM: 612383 MIM: 614312
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CXCL16 PubMed Links

Gene Details

Gene
CXCL16
Gene Name
C-X-C motif chemokine ligand 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001100812.1 282 Intron NP_001094282.1
NM_022059.3 282 Intron NP_071342.2
Gene
MED11
Gene Name
mediator complex subunit 11
There are no transcripts associated with this gene.

Gene
ZMYND15
Gene Name
zinc finger MYND-type containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136046.2 282 Silent Mutation GGA,GGG G72G NP_001129518.1
NM_001267822.1 282 Silent Mutation GGA,GGG G72G NP_001254751.1
NM_032265.2 282 Silent Mutation GGA,GGG G72G NP_115641.1
XM_017025218.1 282 Silent Mutation GGA,GGG G72G XP_016880707.1

View Full Product Details