Product Details

SNP ID
rs139332839
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:42200915 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTCAGGGCTGCGCACTCCACTCT[C/G]GCCAGAACACAAACGGCATTGGCAC
Phenotype
MIM: 608587 MIM: 604260
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GHDC PubMed Links

Gene Details

Gene
GHDC
Gene Name
GH3 domain containing
There are no transcripts associated with this gene.

Gene
STAT5B
Gene Name
signal transducer and activator of transcription 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012448.3 3175 UTR 3 NP_036580.2
XM_005257626.4 3175 Intron XP_005257683.1
XM_017024977.1 3175 UTR 3 XP_016880466.1

View Full Product Details