Product Details

SNP ID
rs140965149
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:75874851 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCACTGCAGGCAGCAGGAGTGGG[C/T]GTTGCGGCCCAGCCGGCCGCGGTCG
Phenotype
MIM: 611041
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM47 PubMed Links

Gene Details

Gene
TRIM47
Gene Name
tripartite motif containing 47
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_033452.2 1576 Missense Mutation ACC,GCC T517A NP_258411.2
XM_005257787.4 1576 Missense Mutation ACC,GCC T279A XP_005257844.1
XM_005257788.4 1576 Missense Mutation ACC,GCC T279A XP_005257845.1
Gene
TRIM65
Gene Name
tripartite motif containing 65
There are no transcripts associated with this gene.

View Full Product Details